Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
MicroKros filter module 40cm 500kD mPES 0,5mm sterile 1 * 1 items
SPECC04-E500-05-S
:
Spectrum Laboratories
MicroKros filter module 40cm 500kD mPES 0,5mm sterile 1 * 1 items
SPECC04-E500-05-S
:
Spectrum Laboratories