Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MidiKros filter module 65cm 10kD mPES 1mm FLL X FLL sterile 1 * 1 items
Catalog # SPECD06-E010-10-S
Supplier:
Spectrum Laboratories
Expand image
MidiKros filter module 65cm 10kD mPES 1mm FLL X FLL sterile 1 * 1 items
Catalog # SPECD06-E010-10-S
Supplier:
Spectrum Laboratories