Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MicroKros filter module 26cm 10kD mPES 1mm sterile 1 * 1 items
Catalog # SPECC04-E010-10-S
Supplier:
Spectrum Laboratories
Expand image
MicroKros filter module 26cm 10kD mPES 1mm sterile 1 * 1 items
Catalog # SPECC04-E010-10-S
Supplier:
Spectrum Laboratories