Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sucrose (Brix) Standard 55% - 1.4308 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS55S
Supplier:
REAGECON
Expand image
Sucrose (Brix) Standard 55% - 1.4308 (± 0.00004), Stabilised, shelf-life of 1 year 1 * 15 mL
Catalog # REGEBS55S
Supplier:
REAGECON