Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Mesna, Reference Standards

Mesna Related Compound B, USP reference standard, Vial, 50 mg
  1392829.
 :   USP
Mesna, Reference Standards
  1392829.
 :  USP
 :  
Mesna, Reference Standards

Mesna Related Compound B, USP reference standard, Vial, 50 mg

 :  
 
1 310,00 €
   
1 * 50 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    USP reference standard
  • Description:
    Mesna Related Compound B
  • Cat. No.:
    1392829.
  • Formula:
    C₄H₁₀O₆S₄
  • Description:
    Mesna Related Compound B
  • Synonym(s):
    2,2'-Disulfanediylbis(ethane-1-sulfonic acid), dipotassium salt, crystal adduct with sodium chloride
  • CAS No.:
    45127-11-5