Order Entry
Finland
Orders LinkContactUsLinkComponent
Laboratory glass cleaner, borer 12 BASIC
Laboratory glass cleaner, borer 12 BASIC
deconex® 12 BASIC
Catalog # DECO500325.25-K05W
Supplier:   Borer Chemie
Laboratory glass cleaner, borer 12 BASIC
Catalog # DECO500325.25-K05W
Supplier:  Borer Chemie
Laboratory glass cleaner, borer 12 BASIC
Laboratory glass cleaner, borer 12 BASIC
deconex® 12 BASIC
Price
Inquire For Price
Log in for availability
1 * 7 kg
Product Details & Documents
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
  • Rinseable without residues and guarantees therefore consistent and error-free results
  • Perfect results even at low concentrations
  • High cleaning performance due to excellent swelling and dispersing capabilities
  • Working solution suitable for multiple use with a broad cleaning spectrum
  • Ideal as an adequate substitute for chromosulphuric acid, not toxic and environmentally friendly
  • EDTA-, NTA- and chlorine-free
Documents
Specifications
  • Description:
    deconex® 12 BASIC
  • Seq:
    03