Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Behenic acid tech. 85%, Technical Grade

Glass bottle for solids, 50 kg
Catalog # A12850.50
Irritant
Behenic acid tech. 85%, Technical Grade
Catalog # A12850.50
CAS Number:  112-85-6
Behenic acid tech. 85%, Technical Grade
Behenic acid tech. 85%, Technical Grade

Glass bottle for solids, 50 kg

CAS Number:  112-85-6
Price
Inquire For Price
Log in for availability
1 * 50 kg
Product Details & Documents

Purity: 85% CAS No.: 112-85-6 Molecular Formula: CH3(CH2)20CO2H Formula Weight: 340.60 Melting Point: 74-78° Boiling Point: 306°/60mm Merck Reference: 14,1023 MDL No.: MFCD00002807

Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:14,1023
  • Boiling Pt:306 °C (60 mmHg)
  • Melting Pt:74…78 °C
  • Density:0.8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.