Order Entry
Finland
Orders LinkContactUsLinkComponent
RoboSep™ Buffer

RoboSep™ Buffer

Buffer containing FBS
Catalog # STMC20104
Marketsource
RoboSep™ Buffer
Catalog # STMC20104
Marketsource
RoboSep™ Buffer
RoboSep™ Buffer

Buffer containing FBS

Price
44,60 €
Log in for availability
1 * 250 mL
Product Details & Documents

RoboSep™ buffer is recommended for EasySep™ cell separation protocols performed by RoboSep™.

Application: Immunology
Species: Human, mouse, non human primate, other, rat

Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications
  • Description:
    Buffer containing FBS
  • Seq:
    01