0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
TISSUE CASSETTE (TC) WHITE 1 * 1 items
LEIH3850400E
:
LEICA HISTOLOGY
TISSUE CASSETTE (TC) WHITE 1 * 1 items
LEIH3850400E
:
LEICA HISTOLOGY