Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EYE WASH DUO 1L 0,9 % NaCl STERILE 1 * 6 items
Catalog # PLUM4804
Supplier:
Plum Safety
Expand image
EYE WASH DUO 1L 0,9 % NaCl STERILE 1 * 6 items
Catalog # PLUM4804
Supplier:
Plum Safety