Order Entry
Finland
ContactUsLinkComponent
Empty Glass GC Column, for Varian(R) 3000, 4000, 6000, and Vista(TM) (FID, Inj. A to Det. A, On-Column), L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 4.0 mm 1 * 1 Each
Catalog # 21722.
Supplier:  Merck
Empty Glass GC Column, for Varian(R) 3000, 4000, 6000, and Vista(TM) (FID, Inj. A to Det. A, On-Column), L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 4.0 mm 1 * 1 Each
Catalog # 21722.
Supplier:  Merck
Frequently Bought Together