Order Entry
Finland
ContactUsLinkComponent
Metal Packed GC Column (General Configuration), phase 3% SP-1500, matrix 80/120 Carbopack B support, L x O.D. x I.D. 10.0 ft (3.0 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12592.
Supplier:  Merck
Metal Packed GC Column (General Configuration), phase 3% SP-1500, matrix 80/120 Carbopack B support, L x O.D. x I.D. 10.0 ft (3.0 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12592.
Supplier:  Merck
Frequently Bought Together