Order Entry
Finland
ContactUsLinkComponent
Metal Packed GC Column (General Configuration), phase 10% SP-2100, matrix 80/100 SUPELCOPORT support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12429.
Supplier:  Merck
Metal Packed GC Column (General Configuration), phase 10% SP-2100, matrix 80/100 SUPELCOPORT support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12429.
Supplier:  Merck
Frequently Bought Together