Order Entry
Finland
ContactUsLinkComponent
Metal Packed GC Column (General Configuration), phase none, matrix 40/60 Carboxen-1000 support, L x O.D. x I.D. 5.0 ft (1.5 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12380.
Supplier:  Merck
Metal Packed GC Column (General Configuration), phase none, matrix 40/60 Carboxen-1000 support, L x O.D. x I.D. 5.0 ft (1.5 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12380.
Supplier:  Merck
Frequently Bought Together