Order Entry
Finland
ContactUsLinkComponent
PTFE Packed GC Column (General Configuration), phase none, matrix Chromosil 330 support, L x O.D. x I.D. 8.0 ft (2.4 m) x 1/8 in. x 2.1 mm, Ends empty, middle 6 ft packed 1 * 1 Each
Catalog # 11496.
Supplier:  Merck
PTFE Packed GC Column (General Configuration), phase none, matrix Chromosil 330 support, L x O.D. x I.D. 8.0 ft (2.4 m) x 1/8 in. x 2.1 mm, Ends empty, middle 6 ft packed 1 * 1 Each
Catalog # 11496.
Supplier:  Merck
Frequently Bought Together