Order Entry
Finland
ContactUsLinkComponent
Carboxen(R)-1010 PLOT Capillary GC Column, L x I.D. 30 m x 0.32 mm, average thickness 15 mum 1 * 1 Each
  24246.
 :  Merck
Carboxen(R)-1010 PLOT Capillary GC Column, L x I.D. 30 m x 0.32 mm, average thickness 15 mum 1 * 1 Each
  24246.
 :  Merck
Frequently Bought Together