Methyl red
Glass bottle, 1 L
Catalog # FFCH62771000
Supplier: FF CHEMICALS
Jump to:
Product Details & Documents
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- CAS Number:493-52-7
- Formula:C₁₅H₁₅N₃O₂
- MDL Number:MFCD00002425
- Molecular Weight:269.3 g/mol
- Merck Index:12,06199
- Melting Pt:178…182 °C
- Density:1.31 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-776-1
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



