Micro Tubes with Thread
Micro tubes without skirted base, neutral cap, 2,0 ml, 1000/case
Catalog # SARS72.693.465
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent with PP screw cap.
- With printed label
- Withstand centrifugation up to 20000×g
- Attached, not screwed-on cap
- Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.
Specifications
- Packed:1000/case
- Capacity:2,0 ml
- Description:Micro tubes without skirted base, neutral cap
- Seq:2