Order Entry
Finland
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 2,10 mm, 100 mm
Catalog # JOURJR-65003NF
Supplier:   VICI JOUR RESEARCH
HPLC columns
Catalog # JOURJR-65003NF
Supplier:  VICI JOUR RESEARCH
HPLC columns

Analytical, 2,10 mm, 100 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
Specifications
  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Length:
    100 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.