Order Entry
Finland
ContactUsLinkComponent
Bar Coded Matrix ScreenMates 96 Well Microplates, V bottom, Polypropylene, Sterile. Ideal for use as a serial dilution plate, daugh ter plate, compound storage plate or for non-binding microtiter assays. V bottom shape is ideal for low volume aspirat 1 * 80 items
Catalog # MATRBC30182
Bar Coded Matrix ScreenMates 96 Well Microplates, V bottom, Polypropylene, Sterile. Ideal for use as a serial dilution plate, daugh ter plate, compound storage plate or for non-binding microtiter assays. V bottom shape is ideal for low volume aspirat 1 * 80 items
Catalog # MATRBC30182
Frequently Bought Together