Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLUTION PH 7.0 HAEMATOLOGY 5L 1 * 5 L
Catalog # RALA330370-5000
Supplier:
RAL
Expand image
BUFFER SOLUTION PH 7.0 HAEMATOLOGY 5L 1 * 5 L
Catalog # RALA330370-5000
Supplier:
RAL