Order Entry
Finland
Orders LinkContactUsLinkComponent
Initial calibration standards

Initial calibration standards

Initial calibration verification standard 2, (2% HNO3/tr HF) 125 ml, 10 mg/l: Sn, Ti 125 ml, 2% HNO₃, 1
Catalog # PERCN9303826
Supplier:   PerkinElmer
Initial calibration standards
Catalog # PERCN9303826
Supplier:  PerkinElmer
CAS Number:  
Initial calibration standards

Initial calibration verification standard 2, (2% HNO3/tr HF) 125 ml, 10 mg/l: Sn, Ti 125 ml, 2% HNO₃, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications

Specifications

  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Description:
    Initial calibration verification standard 2, (2% HNO<sub>3</sub>/tr HF) 125 ml, 10 mg/l: Sn, Ti 125 ml

Specifications