Order Entry
Finland
Orders LinkContactUsLinkComponent
Initial calibration standards

Initial calibration standards

Initial calibration verification standard 1, (5% HNO3/tr Tartaric Acid), 125 ml, 1000 mg/l: Ca, Fe, K, Mg, Na, Sr and 10 mg/l: Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Tl, V, Zn, Th, U, 5% HNO₃, 1
Catalog # PERCN9303825
Supplier:   PerkinElmer
Initial calibration standards
Catalog # PERCN9303825
Supplier:  PerkinElmer
CAS Number:  
Initial calibration standards

Initial calibration verification standard 1, (5% HNO3/tr Tartaric Acid), 125 ml, 1000 mg/l: Ca, Fe, K, Mg, Na, Sr and 10 mg/l: Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Tl, V, Zn, Th, U, 5% HNO₃, 1

CAS Number:  
Price
826,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    1
  • Description:
    Initial calibration verification standard 1, (5% HNO<sub>3</sub>/tr Tartaric Acid), 125 ml, 1000 mg/l: Ca, Fe, K, Mg, Na, Sr and 10 mg/l: Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Tl, V, Zn, Th, U

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.