Order Entry
Finland
Orders LinkContactUsLinkComponent
Cleanroom overalls, Kimtech™ A5

Cleanroom overalls, Kimtech™ A5

Without hood, White, 4XL, +
Catalog # 113-1219
   Waste Reducing
Supplier:   Ansell
Cleanroom overalls, Kimtech™ A5
Catalog # 113-1219
Supplier:  Ansell
   Waste Reducing
Cleanroom overalls, Kimtech™ A5
Cleanroom overalls, Kimtech™ A5

Without hood, White, 4XL, +

Price
581,00 €
Log in for availability
1 * 25 Items
Product Details & Documents

Kimtech™ A5 sterile cleanroom apparel with CLEAN-DON™ technology is made of SMS (Spunbond Melt-blown Spunbond) material. It features built-in snaps that gather up legs and arms to aid aseptic gowning. Garments have a thumb loop that prevents the sleeve from rolling back and exposing arm or wrist when in use.

  • Cloth-like comfort
  • Innovative inside-out fold pattern - reduces the risk of touching and contaminating the outside of the apparel when opening or donning
  • Highly visible blue line along the inside of the garment signals the proper place to grasp while gowning
  • Vacuum packaging reduces space, so more products fit into gowning area storage

Suitable for use in Class 100, ISO 5, 3.5 environments.

Colour: White

Certifications: PPE Cat III (EU) 2016/425, Type 5/6

Packaging: Vacuum packed and double-bagged for added sterility assurance.

Specifications
  • Clothing Size:
    4XL
  • Sterile:
    +
  • Colour:
    White
  • Description:
    Without hood
  • Seq:
    07
  • Cat. No.:
    113-1219
  • Cleanroom class:
    100/ISO 5
  • Garment application:
    Cleanroom|Laboratory
  • Environmentally Preferable:
    Waste Reducing
Complementary Products

Product Information

  • Catalog #
    Description
    Shoe size
    Sterile
    Colour
    Length
    Availability
    Price
  • 113-8197
    Wrap-around vinyl foot
    S/M
    +
    White
    Log in for availability
    1 * 100 Pari - 832,00 €

  • 113-1227
    Wrap-around vinyl foot
    U*
    +
    White
    Log in for availability
    1 * 100 Pari - 832,00 €

  • 113-7736
    Standard vinyl sole
    U*
    +
    White
    Log in for availability
    1 * 100 Pari - 1 040,00 €

  • 113-7735
    Standard vinyl sole
    S/M
    +
    White
    Log in for availability
    1 * 100 Pari - 1 040,00 €

  • 113-7733
    457 mm
    +
    White
    Log in for availability
    1 * 100 Pari - 520,00 €

  • 113-7732
    With integrated mask
    +
    01
    Log in for availability
    1 * 75 Items - 754,00 €

  • 114-9642
    Standard
    +
    03
    Log in for availability
    1 * 100 Items - 471,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.