Order Entry
Finland
Orders LinkContactUsLinkComponent
Samplers, Liquid-Sampler

Samplers, Liquid-Sampler

Liquid-Sampler, place onto bottle, 32×455 mm, 50 ml
Catalog # 331-0034
Supplier:   BURKLE
Samplers, Liquid-Sampler
Catalog # 331-0034
Supplier:  BURKLE
Samplers, Liquid-Sampler
Samplers, Liquid-Sampler

Liquid-Sampler, place onto bottle, 32×455 mm, 50 ml

Price
430,00 €
Log in for availability
1 * 1 Items
Product Details & Documents

Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.

  • Easy one handed operation
  • Discharge funnel for safe filling of sample bottles

Documents

Specifications
  • Capacity:
    50 ml
  • Description:
    Liquid-Sampler, place onto bottle, 32×455 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.