Order Entry
Finland
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ MabSelect™

Affinity chromatography columns, HiTrap™ MabSelect™

HiTrap™ MabSelect™, PP, 85 µm, 7×25 mm, 5×1 ml, <4 ml/min
Catalog # 28-4082-53
Supplier:   Cytiva
Affinity chromatography columns, HiTrap™ MabSelect™
Catalog # 28-4082-53
Supplier:  Cytiva
Affinity chromatography columns, HiTrap™ MabSelect™
Affinity chromatography columns, HiTrap™ MabSelect™

HiTrap™ MabSelect™, PP, 85 µm, 7×25 mm, 5×1 ml, <4 ml/min

Price
1 090,00 €
Log in for availability
1 * 5 Items
Product Details & Documents

HiTrap™ MabSelect™ columns are prepacked with MabSelect™, a Bioprocess medium for capuring of monoclonal antibodies(Mabs) from large sample volumes.

  • Specificity of binding to the Fc region of IgG is similar to that of native protein A, which allows one-step purification
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
Specifications
  • Flow rate:
    <4 ml/min
  • Particle Size:
    85 µm
  • Capacity:
    5×1 ml
  • Description:
    HiTrap™ MabSelect™
  • Seq:
    01
  • Column hardware:
    PP
  • Bed dimensions:
    7×25 mm
  • Cat. No.:
    28-4082-53
  • Max. pressure:
    5 bar
  • Storage:
    2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
  • pH range:
    3 - 10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.