Order Entry
Finland
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ MabSelect Xtra™

Affinity chromatography columns, HiTrap™ MabSelect Xtra™

HiTrap™ MabSelect Xtra™, PP, 75 µm, 16×25 mm, 1×5 ml, <20 ml/min
Catalog # 28-4082-60
Supplier:   Cytiva
Affinity chromatography columns, HiTrap™ MabSelect Xtra™
Catalog # 28-4082-60
Supplier:  Cytiva
Affinity chromatography columns, HiTrap™ MabSelect Xtra™
Affinity chromatography columns, HiTrap™ MabSelect Xtra™

HiTrap™ MabSelect Xtra™, PP, 75 µm, 16×25 mm, 1×5 ml, <20 ml/min

Price
1 050,00 €
Log in for availability
1 * 1 Items
Product Details & Documents

HiTrap™ MabSelect Xtra™ columns are prepacked with MabSelect Xtra™, a BioProcess medium for capture of monoclonal antbodies from large sample volumes.

  • Increased dynamic binding capacity with smaller particle size and greater porosity than MabSelect™ medium
  • MabSelect™ Xtra uses the same recombinant protein A ligand as MabSelect™
  • Oriented coupling of recombinant protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification

The increased capacity of the medium is well suited for purification of Mabs from high-level expression feedstock.

Specifications
  • Flow rate:
    <20 ml/min
  • Particle Size:
    75 µm
  • Capacity:
    1×5 ml
  • Description:
    HiTrap™ MabSelect Xtra™
  • Seq:
    02
  • Column hardware:
    PP
  • Bed dimensions:
    16×25 mm
  • Cat. No.:
    28-4082-60
  • Max. pressure:
    5 bar
  • pH range:
    3 - 10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.