Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Samplers, Liquid-Sampler

Liquid-Sampler, place onto bottle, 32×535 mm, 100 ml
  331-0035
 :   BURKLE
Samplers, Liquid-Sampler
  331-0035
 :  BURKLE
Samplers, Liquid-Sampler
Samplers, Liquid-Sampler

Liquid-Sampler, place onto bottle, 32×535 mm, 100 ml

 
496,00 €
   
1 * 1 Items
 

Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.

  • Easy one handed operation
  • Discharge funnel for safe filling of sample bottles
 
  • Capacity:
    100 ml
  • Description:
    Liquid-Sampler, place onto bottle, 32×535 mm