Order Entry
Finland
Orders LinkContactUsLinkComponent
Accessories for Cubis® balances
Accessories for Cubis® balances
Display cable, 3 m, for MSA and MSU models, for remote setup of display and weighing unit
Catalog # SARBYCC01-MSD3
Supplier:   Sartorius Balances
Accessories for Cubis® balances
Catalog # SARBYCC01-MSD3
Supplier:  Sartorius Balances
Accessories for Cubis® balances
Display cable, 3 m, for MSA and MSU models, for remote setup of display and weighing unit
Price
232,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Display cable, 3 m, for MSA and MSU models, for remote setup of display and weighing unit
  • Seq:
    13