Order Entry
Finland
Orders LinkContactUsLinkComponent
Replacement Erlenmeyer flask caps, Corning®

Replacement Erlenmeyer flask caps, Corning®

Flat cap, 70 mm, for 3 L flask
Catalog # 734-4194
Supplier:   Corning
Replacement Erlenmeyer flask caps, Corning®
Catalog # 734-4194
Supplier:  Corning
Replacement Erlenmeyer flask caps, Corning®
Replacement Erlenmeyer flask caps, Corning®

Flat cap, 70 mm, for 3 L flask

Price
191,00 €
Log in for availability
1 * 24 Items
Product Details & Documents

Made of PP.

Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.

Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Specifications
  • Description:
    Flat cap, 70 mm, for 3 L flask
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.