Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CAPPING REAGENT B2 1L 60% LUTIDINE IN ACN 1 * 500 mL
Catalog # 8.57006.0500
Supplier:
Novabiochem (Part of Merck)
Certificates
Expand image
CAPPING REAGENT B2 1L 60% LUTIDINE IN ACN 1 * 500 mL
Catalog # 8.57006.0500
Supplier:
Novabiochem (Part of Merck)
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation