Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH BUFFER KIT PH 4 ,7 , 10 1 * 1 KIT
Catalog # YSIA603824
Supplier:
YSI ENVIRONMENTAL
Expand image
PH BUFFER KIT PH 4 ,7 , 10 1 * 1 KIT
Catalog # YSIA603824
Supplier:
YSI ENVIRONMENTAL