Order Entry
Finland
Orders LinkContactUsLinkComponent
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%

3,5-Dimethyl-4-hydroxybenzonitrile ≥98%

Glass bottle for solids, 25 g
Catalog # H26872.14
Toxic
Irritant
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%
Catalog # H26872.14
CAS Number:  4198-90-7
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%

Glass bottle for solids, 25 g

CAS Number:  4198-90-7
Price
730,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 4198-90-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 123-127° MDL No.: MFCD00221716

Synonyms:  4-Hydroxy-3,5-dimethylbenzonitrile
  • CAS Number:4198-90-7
  • Formula:C₉H₉NO
  • MDL Number:MFCD00221716
  • Molecular Weight:147.18 g/mol
  • Melting Pt:123…127 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.