Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TARGET MICROVIAL SOLID GLASS 100UL 1 * 12 items
Catalog # NSCAC4010-12
Supplier:
THERMO NATIONAL SCIENTIFIC
Expand image
TARGET MICROVIAL SOLID GLASS 100UL 1 * 12 items
Catalog # NSCAC4010-12
Supplier:
THERMO NATIONAL SCIENTIFIC