n-Hexadecane, Supelco®
Glass bottle, 25 mL
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Hexadecane, Cetane
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
- Boiling Pt:287 °C (1013 hPa)
- Melting Pt:18 °C
- Density:0.77331 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:135 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:208-878-9
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



