Order Entry
Finland
Orders LinkContactUsLinkComponent
Polyethylene Glycol, Reference Standards

Polyethylene Glycol, Reference Standards

Polyethylene Glycol 3000, USP reference standard, Vial, 1 g
Tuotenumero 1546525.
Toimittaja:   USP
Polyethylene Glycol, Reference Standards
Tuotenumero 1546525.
Toimittaja:  USP
CAS numero:  
Polyethylene Glycol, Reference Standards

Polyethylene Glycol 3000, USP reference standard, Vial, 1 g

Siirry kohtaan:
CAS numero:  
Price
420,00 €
Kirjaudu sisään saatavuuden tarkistamiseksi
1 * 1 g
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Tekniset tiedot

Tekniset tiedot

  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • Description:
    Polyethylene Glycol 3000
  • Cat. No.:
    1546525.
  • Description:
    Polyethylene Glycol
  • Synonym(s):
    PEG
  • CAS No.:
    25322-68-3

Tekniset tiedot