Nitromethane 99+% for analysis
100 mL
Supplier: Thermo Fisher Scientific
Nitromethane 99+% for analysis
Catalog # ACRO213371000
Supplier: Thermo Fisher Scientific
CAS Number: 75-52-5
Jump to:
Product Details & Documents
- CAS Number:75-52-5
- Formula:CH₃NO₂
- MDL Number:MFCD00007400
- Molecular Weight:61.04 g/mol
- Merck Index:13,06644
- Boiling Pt:100…102 °C (760 mmHg)
- Melting Pt:–29 °C
- Density:1.12 g/cm³
- Flash Pt:35 °C
- Storage Temperature:Ambient
- EINECS:200-876-6
- UN:1261
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3805 to 1.3825 (20°C, 589 nm)
- Water:≤0.5 % (Coulometric)
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.1 ppm
Specifications
- Pk:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



