Order Entry
Finland
Orders LinkContactUsLinkComponent
Potassium bromate ≥99%

Potassium bromate ≥99%

Glass bottle for solids, 500 g
Catalog # A18258.36
Oxidizing
Toxic
Health
Potassium bromate ≥99%
Catalog # A18258.36
CAS Number:  7758-01-2
Potassium bromate ≥99%
Potassium bromate ≥99%

Glass bottle for solids, 500 g

CAS Number:  7758-01-2
Price
126,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 7758-01-2 Molecular Formula: KBrO3 Formula Weight: 167.00 Odour: Odourless Melting Point: ca 370° dec. UN No.: 1484 MDL No.: MFCD00011359

  • CAS Number:7758-01-2
  • Formula:KBrO₃
  • MDL Number:MFCD00011359
  • Molecular Weight:167 g/mol
  • Merck Index:14,7617
  • Melting Pt:∼ 370 °C (dec.)
  • Density:3.27
  • Storage Temperature:Ambient
  • EINECS:231-829-8
  • UN:1484
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.