L(-)-Mannose ≥99%
Glass bottle for solids, 0,25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 10030-80-5 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 129-131° Beilstein Reference: 1724628
Synonyms: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose
- CAS Number:10030-80-5
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00136021
- Molecular Weight:180.16 g/mol
- Boiling Pt:376 °C (1013 hPa)
- Melting Pt:129…131 °C
- Storage Temperature:Ambient
- EINECS:233-080-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



