Order Entry
Finland
Orders LinkContactUsLinkComponent
 

(±)-2-Octanol ≥98%

Glass bottle, 100 mL
Catalog # A16985.AE
Irritant
(±)-2-Octanol ≥98%
Catalog # A16985.AE
CAS Number:  123-96-6
(±)-2-Octanol ≥98%
(±)-2-Octanol ≥98%

Glass bottle, 100 mL

CAS Number:  123-96-6
Price
54,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 98% CAS No.: 123-96-6 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 76°(169°F) Refractive Index: 1.4260 Odour: Pleasant Melting Point: -38° Boiling Point: 179-180° Beilstein Reference: 1719325 Merck Reference: 14,6752 MDL No.: MFCD00004591

Synonyms:  (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
  • CAS Number:123-96-6
  • Formula:C₈H₁₈O
  • MDL Number:MFCD00004591
  • Molecular Weight:130.23 g/mol
  • Merck Index:14,6752
  • Boiling Pt:179…180 °C
  • Melting Pt:–38 °C
  • Density:0.819
  • Flash Pt:76 °C (169 °F)
  • Storage Temperature:Ambient
  • EINECS:204-667-0
  • UN:1987
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications