Order Entry
Finland
Orders LinkContactUsLinkComponent
(±)-2-Octanol ≥98%

(±)-2-Octanol ≥98%

Glass bottle, 2.500 mL
Catalog # A16985.0F
Irritant
(±)-2-Octanol ≥98%
Catalog # A16985.0F
CAS Number:  123-96-6
(±)-2-Octanol ≥98%
(±)-2-Octanol ≥98%

Glass bottle, 2.500 mL

CAS Number:  123-96-6
Price
188,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 98% CAS No.: 123-96-6 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 76°(169°F) Refractive Index: 1.4260 Odour: Pleasant Melting Point: -38° Boiling Point: 179-180° Beilstein Reference: 1719325 Merck Reference: 14,6752 MDL No.: MFCD00004591

Synonyms:  (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
  • CAS Number:123-96-6
  • Formula:C₈H₁₈O
  • MDL Number:MFCD00004591
  • Molecular Weight:130.23 g/mol
  • Merck Index:14,6752
  • Boiling Pt:179…180 °C
  • Melting Pt:–38 °C
  • Density:0.819
  • Flash Pt:76 °C (169 °F)
  • Storage Temperature:Ambient
  • EINECS:204-667-0
  • UN:1987
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.