Order Entry
Finland
Orders LinkContactUsLinkComponent
 

4'-Fluoroacetophenone ≥99%

Glass bottle for solids, 100 g
Catalog # A13950.22
Irritant
4'-Fluoroacetophenone ≥99%
Catalog # A13950.22
CAS Number:  403-42-9
4'-Fluoroacetophenone ≥99%
4'-Fluoroacetophenone ≥99%

Glass bottle for solids, 100 g

CAS Number:  403-42-9
Price
162,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 403-42-9 Molecular Formula: C8H7FO Formula Weight: 138.14 Flash Point: 71°(159°F) Refractive Index: 1.5110 Melting Point: 4° Boiling Point: 194-196° Beilstein Reference: 386013 MDL No.: MFCD00000354

Synonyms:  1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
  • CAS Number:403-42-9
  • Formula:C₈H₇FO
  • MDL Number:MFCD00000354
  • Molecular Weight:138.14 g/mol
  • Boiling Pt:194…196 °C
  • Melting Pt:4 °C
  • Density:1.141
  • Flash Pt:71 °C (159 °F)
  • Storage Temperature:Ambient
  • EINECS:206-960-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications