4'-Fluoroacetophenone ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
4'-Fluoroacetophenone ≥99%
Catalog # A13950.14
Supplier: Thermo Fisher Scientific
CAS Number: 403-42-9
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 403-42-9 Molecular Formula: C8H7FO Formula Weight: 138.14 Flash Point: 71°(159°F) Refractive Index: 1.5110 Melting Point: 4° Boiling Point: 194-196° Beilstein Reference: 386013 MDL No.: MFCD00000354
Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
- CAS Number:403-42-9
- Formula:C₈H₇FO
- MDL Number:MFCD00000354
- Molecular Weight:138.14 g/mol
- Boiling Pt:194…196 °C
- Melting Pt:4 °C
- Density:1.141
- Flash Pt:71 °C (159 °F)
- Storage Temperature:Ambient
- EINECS:206-960-9
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



