Order Entry
Finland
Orders LinkContactUsLinkComponent
4-Hydroxybenzaldehyde ≥98%

4-Hydroxybenzaldehyde ≥98%

Glass bottle for solids, 500 g
Catalog # A13580.36
Irritant
4-Hydroxybenzaldehyde ≥98%
Catalog # A13580.36
CAS Number:  123-08-0
4-Hydroxybenzaldehyde ≥98%
4-Hydroxybenzaldehyde ≥98%

Glass bottle for solids, 500 g

CAS Number:  123-08-0
Price
322,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 123-08-0 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 174°(345°F) Melting Point: 115-119° Boiling Point: 191°/50mm Beilstein Reference: 471352 Merck Reference: 14,4811 MDL No.: MFCD00006939

  • CAS Number:123-08-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00006939
  • Molecular Weight:122.12 g/mol
  • Merck Index:14,4811
  • Boiling Pt:191 °C (50 mmHg)
  • Melting Pt:115…119 °C
  • Density:1.129
  • Flash Pt:174 °C (345 °F)
  • Storage Temperature:Ambient
  • EINECS:204-599-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.