Order Entry
Finland
Orders LinkContactUsLinkComponent
 

4-Hydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # A13580.14
Irritant
4-Hydroxybenzaldehyde ≥98%
Catalog # A13580.14
CAS Number:  123-08-0
4-Hydroxybenzaldehyde ≥98%
4-Hydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  123-08-0
Price
44,40 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 123-08-0 Molecular Formula: C7H6O2 Formula Weight: 122.12 Flash Point: 174°(345°F) Melting Point: 115-119° Boiling Point: 191°/50mm Beilstein Reference: 471352 Merck Reference: 14,4811 MDL No.: MFCD00006939

  • CAS Number:123-08-0
  • Formula:C₇H₆O₂
  • MDL Number:MFCD00006939
  • Molecular Weight:122.12 g/mol
  • Merck Index:14,4811
  • Boiling Pt:191 °C (50 mmHg)
  • Melting Pt:115…119 °C
  • Density:1.129
  • Flash Pt:174 °C (345 °F)
  • Storage Temperature:Ambient
  • EINECS:204-599-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications