Order Entry
Finland
Orders LinkContactUsLinkComponent
 

2,5-Dichlorophenol ≥98%

Glass bottle for solids, 100 g
Catalog # B24787.22
Irritant
Environment
2,5-Dichlorophenol ≥98%
Catalog # B24787.22
CAS Number:  583-78-8
2,5-Dichlorophenol ≥98%
2,5-Dichlorophenol ≥98%

Glass bottle for solids, 100 g

CAS Number:  583-78-8
Price
125,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 583-78-8 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 55-58° Boiling Point: 211° Beilstein Reference: 1907692 UN No.: 2020 MDL No.: MFCD00002174

  • CAS Number:583-78-8
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002174
  • Molecular Weight:163 g/mol
  • Boiling Pt:211 °C
  • Melting Pt:55…58 °C
  • Storage Temperature:Ambient
  • EINECS:209-520-4
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.