Order Entry
Finland
Orders LinkContactUsLinkComponent
 

2,5-Dichlorophenol ≥98%

Glass bottle for solids, 25 g
Catalog # B24787.14
Irritant
Environment
2,5-Dichlorophenol ≥98%
Catalog # B24787.14
CAS Number:  583-78-8
2,5-Dichlorophenol ≥98%
2,5-Dichlorophenol ≥98%

Glass bottle for solids, 25 g

CAS Number:  583-78-8
Price
50,40 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 583-78-8 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 55-58° Boiling Point: 211° Beilstein Reference: 1907692 UN No.: 2020 MDL No.: MFCD00002174

  • CAS Number:583-78-8
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002174
  • Molecular Weight:163 g/mol
  • Boiling Pt:211 °C
  • Melting Pt:55…58 °C
  • Storage Temperature:Ambient
  • EINECS:209-520-4
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications