4'-Cyclohexylacetophenone ≥99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
4'-Cyclohexylacetophenone ≥99%
Catalog # B20838.06
Supplier: Thermo Fisher Scientific
CAS Number: 18594-05-3
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 18594-05-3 Molecular Formula: C14H18O Formula Weight: 202.30 Melting Point: 66-70° Boiling Point: 179-181°/14mm MDL No.: MFCD00001453
Synonyms: p-Cyclohexylacetophenone
- CAS Number:18594-05-3
- Formula:C₁₄H₁₈O
- MDL Number:MFCD00001453
- Molecular Weight:202.3 g/mol
- Boiling Pt:179…181 °C (14 mmHg)
- Melting Pt:66…70 °C
- Storage Temperature:Ambient
- EINECS:242-432-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



