Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Behenic acid tech. 85%, Technical Grade

Glass bottle for solids, 250 g
Catalog # A12850.30
Irritant
Behenic acid tech. 85%, Technical Grade
Catalog # A12850.30
CAS Number:  112-85-6
Behenic acid tech. 85%, Technical Grade
Behenic acid tech. 85%, Technical Grade

Glass bottle for solids, 250 g

CAS Number:  112-85-6
Price
81,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 85% CAS No.: 112-85-6 Molecular Formula: CH3(CH2)20CO2H Formula Weight: 340.60 Melting Point: 74-78° Boiling Point: 306°/60mm Merck Reference: 14,1023 MDL No.: MFCD00002807

Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:14,1023
  • Boiling Pt:306 °C (60 mmHg)
  • Melting Pt:74…78 °C
  • Density:0.8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications