Order Entry
Finland
Orders LinkContactUsLinkComponent
3,4-Dichlorophenol ≥99%

3,4-Dichlorophenol ≥99%

Glass bottle for solids, 100 g
Catalog # A12256.22
Irritant
3,4-Dichlorophenol ≥99%
Catalog # A12256.22
CAS Number:  95-77-2
3,4-Dichlorophenol ≥99%
3,4-Dichlorophenol ≥99%

Glass bottle for solids, 100 g

CAS Number:  95-77-2
Price
462,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 95-77-2 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 64-68° Boiling Point: 252-255° Beilstein Reference: 1907693 UN No.: 2020 MDL No.: MFCD00002258

  • CAS Number:95-77-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002258
  • Molecular Weight:163 g/mol
  • Boiling Pt:252…255 °C
  • Melting Pt:64…68 °C
  • Density:1.6 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-450-5
  • UN:2020
  • ADR:6.1,III
  •  :Density: 1.6 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications